2-(3-Bromphenyl)-5-methyl-1,3,4-oxadiazol

Lieferant: COMBI-BLOCKS

HC-6376-5G HC-6376-1G
COBBHC-6376-5GEA 362 EUR
COBBHC-6376-5G COBBHC-6376-1G
2-(3-Bromphenyl)-5-methyl-1,3,4-oxadiazol
2-(3-Bromophenyl)-5-methyl-1,3,4-oxadiazole
Formel: C₉H₇BrN₂O
Molecular Weight: 239,0700 g/mol
MDL: MFCD08691468
CAS-Nummer: 41491-53-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR