2-(3-Bromphenyl)-5-propyl-1,3,4-oxadiazol

Lieferant: COMBI-BLOCKS

HC-6401-5G HC-6401-1G
COBBHC-6401-5GEA 595 EUR
COBBHC-6401-5G COBBHC-6401-1G
2-(3-Bromphenyl)-5-propyl-1,3,4-oxadiazol
2-(3-Bromophenyl)-5-propyl-1,3,4-oxadiazole
Formel: C₁₁H₁₁BrN₂O
MDL: MFCD09878393
CAS-Nummer: 957065-95-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR