1-Methylcyclopropancarbonsäure
Lieferant: CHEMPUR
|
Gefahr
|
Synonyme:
1-methylcyclopropane-1-carboxylic acid
JFD02330.250
JFD02330.1
CHMPJFD02330.250EA
0
EUR
CHMPJFD02330.250
MAYBJFD02330.1
1-Methylcyclopropancarbonsäure
1-Methylcyclopropanecarboxylic acid
Formel:
C₅H₈O₂ Molecular Weight: 100.12 g/mol |
MDL:
MFCD00001290 CAS-Nummer: 6914-76-7 EINECS: 230-020-7 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...