2-Decin-1-ol

Lieferant: FLUOROCHEM

009119-10G 009119-50G
FLUO009119-10GEA 0 EUR
FLUO009119-10G FLUO009119-50G
2-Decin-1-ol
2-Decin-1-ol
Formel: C₁₀H₁₈O
MDL: MFCD00041567
CAS-Nummer: 4117-14-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR