(±)-Ethyl-2-bromvalerat ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Ethyl 2-bromovalerate
B25530.14
B25530.22
B25530.14EA
29.2
EUR
B25530.14
B25530.22
(±)-Ethyl-2-bromvalerat ≥97%
(±)-Ethyl-2-bromvalerat
Purity: 97%
CAS No.: 615-83-8
Molecular Formula: C7H13BrO2
Formula Weight: 209.09
Flash Point: 77°(170°F)
Refractive Index: 1.4480
Boiling Point: 190-192°
Beilstein Reference: 1098901
UN No.: 3265
MDL No.: MFCD00000159
Formel:
C₇H₁₃BrO₂ Molecular Weight: 209.09 g/mol Siedepunkt: 190…192 °C Dichte: 1.226 Flammpunkt: 77 °C (170 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00000159 CAS-Nummer: 615-83-8 EINECS: 210-450-1 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...