DL-β-Leucin

Lieferant: BIONET RESEARCH

Synonyme: 3-Amino-4-methylpentanoic acid

7T-0026.500 7T-0026.1 7T-0026.5 7T-0026.10
BIOT7T-0026.500EA 0 EUR
BIOT7T-0026.500 BIOT7T-0026.1 BIOT7T-0026.5 BIOT7T-0026.10
DL-β-Leucin
DL-β-Leucin
Formel: C₆H₁₃NO₂
Molecular Weight: 131.1729 g/mol
MDL: MFCD00800505
CAS-Nummer: 5699-54-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR