3-Amino-2-methylbenzotrifluorid

Lieferant: FLUOROCHEM

Synonyme: 2-Methyl-3-(trifluoromethyl)aniline

006839-1G 006839-5G 006839-25G
FLUO006839-1GEA 31.1 EUR
FLUO006839-1G FLUO006839-5G FLUO006839-25G
3-Amino-2-methylbenzotrifluorid
3-Amino-2-methylbenzotrifluorid
Formel: C₈H₈F₃N
Molecular Weight: 175.16 g/mol
MDL: MFCD00153216
CAS-Nummer: 54396-44-0
EINECS: 259-145-5
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR