5-Amino-2-methylbenzotrifluorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Methyl-3-(trifluoromethyl)aniline

M-Clarity™ Qualitätsstufe = MQ100

559423-1G 559423-5G
SIAL559423-1GEA 34.4 EUR
SIAL559423-1G SIAL559423-5G
5-Amino-2-methylbenzotrifluorid, Sigma-Aldrich®
5-Amino-2-methylbenzotrifluorid
Formel: C₈H₈F₃N
Molecular Weight: 175,16 g/mol
MDL: MFCD01631582
CAS-Nummer: 65934-74-9
UN: 2810
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR