4'-Chlor-2'-methylacetanilid

Lieferant: Thermo Scientific

Synonyme: N-(4-Chloro-2-methylphenyl)acetamide

TL00898.10 TL00898.50
MAYBTL00898.10EA 0 EUR
MAYBTL00898.10 MAYBTL00898.50
4'-Chlor-2'-methylacetanilid
4'-Chlor-2'-methylacetanilid
Formel: C₉H₁₀ClNO
Molecular Weight: 183,64 g/mol
MDL: MFCD00027399
CAS-Nummer: 5202-86-8
EINECS: 225-991-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR