Monoethylsuccinat 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Ethyl hydrogen succinate, 4-Ethoxy-4-oxobutanoic acid, mono-Ethyl succinate
H55116.03
H55116.06
H55116.14
H55116.03EA
32
EUR
H55116.03
H55116.06
H55116.14
Monoethylsuccinat 95%
Monoethylsuccinat
Purity: 95%
CAS No.: 1070-34-4
Molecular Formula: C6H10O4
Formula Weight: 146.14
Flash Point: 113°(235°F)
Refractive Index: 1.4320
Boiling Point: 248°
MDL No.: MFCD00053346
Formel:
C₆H₁₀O₄ Molecular Weight: 146,14 g/mol Siedepunkt: 248 °C Dichte: 1,141 Flammpunkt: 113 °C (235 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00053346 CAS-Nummer: 1070-34-4 EINECS: 213-973-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...