Monomethylsuccinat

Lieferant: Thermo Scientific

Synonyme: 4-Methoxy-4-oxobutanoic acid, Methyl hydrogen succinate, Succinic acid monomethyl ester

BTB06937.1 BTB06937.10
MAYBBTB06937.1EA 0 EUR
MAYBBTB06937.1 MAYBBTB06937.10
Monomethylsuccinat
Monomethylsuccinat
Formel: C₅H₈O₄
Molecular Weight: 132,11 g/mol
MDL: MFCD00002788
CAS-Nummer: 3878-55-5
EINECS: 223-408-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR