Monomethylsuccinat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
4-Methoxy-4-oxobutanoic acid, Methyl hydrogen succinate, Succinic acid monomethyl ester
M-Clarity™ Qualitätsstufe = MQ200
M81101-100G
M81101-25G
SIALM81101-100GEA
137
EUR
SIALM81101-100G
SIALM81101-25G
Monomethylsuccinat, Sigma-Aldrich®
Monomethylsuccinat
Formel:
C₅H₈O₄ Molecular Weight: 132,11 g/mol |
MDL:
MFCD00002788 CAS-Nummer: 3878-55-5 EINECS: 223-408-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...