2-Brom-4-nitro(trifluormethoxy)benzol ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-Bromo-4-nitro-1-(trifluoromethoxy)benzene
H26173.03
H26173.06
H26173.03EA
33.9
EUR
H26173.03
H26173.06
2-Brom-4-nitro(trifluormethoxy)benzol ≥98%
2-Brom-4-nitro(trifluormethoxy)benzol
Purity: 98%
CAS No.: 200958-40-3
Molecular Formula: C7H3BrF3NO3
Formula Weight: 286.00
Refractive Index: 1.508
Formel:
C₇H₃BrF₃NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD04973758 CAS-Nummer: 200958-40-3 EINECS: None |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...