2-Brom-4-nitro(trifluormethoxy)benzol

Lieferant: CHEMPUR

Synonyme: 2-Bromo-4-nitro-1-(trifluoromethoxy)benzene

FL13788.POR 013788-25G 013788-5G
CHMPFL13788.POREA 0 EUR
CHMPFL13788.POR FLUO013788-25G FLUO013788-5G
2-Brom-4-nitro(trifluormethoxy)benzol
2-Brom-4-nitro(trifluormethoxy)benzol
Formel: C₇H₃BrF₃NO₃
Molecular Weight: 286,00 g/mol
MDL: MFCD04973758
CAS-Nummer: 200958-40-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR