Dimethylcyclobutan-1,1-dicarboxylat

Lieferant: COMBI-BLOCKS

Synonyme: Dimethyl cyclobutane-1,1-dicarboxylate, Dimethyl 1,1-cyclobutanedicarboxylate

OT-0564-100G OT-0564-5G OT-0564-25G
COBBOT-0564-100GEA 595 EUR
COBBOT-0564-100G COBBOT-0564-5G COBBOT-0564-25G
Dimethylcyclobutan-1,1-dicarboxylat
Dimethylcyclobutan-1,1-dicarboxylat
Formel: C₈H₁₂O₄
Molecular Weight: 172.180 g/mol
MDL: MFCD00082719
CAS-Nummer: 10224-72-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR