1-Ethyl-1-methylpyrrolidiniumbis(trifluormethylsulfonyl)imid, Sigma-Aldrich®
Lieferant: Merck
713155-50G
713155-5G
SIAL713155-50GEA
449
EUR
SIAL713155-50G
SIAL713155-5G
1-Ethyl-1-methylpyrrolidiniumbis(trifluormethylsulfonyl)imid, Sigma-Aldrich®
1-Ethyl-1-methylpyrrolidiniumbis(trifluormethylsulfonyl)imid
Formel:
C₉H₁₆F₆N₂O₄S₂ |
CAS-Nummer:
223436-99-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...