trans-4-Pentylcyclohexancarbonsäure
Lieferant: COMBI-BLOCKS
CA-4501-5G
CA-4501-25G
CA-4501-100G
CA-4501-1G
COBBCA-4501-5GEA
31.2
EUR
COBBCA-4501-5G
COBBCA-4501-25G
COBBCA-4501-100G
COBBCA-4501-1G
trans-4-Pentylcyclohexancarbonsäure
trans-4-Pentylcyclohexanecarboxylic acid
Formel:
C₁₁H₂₀O₂ Molecular Weight: 198.30 g/mol |
MDL:
MFCD00009964 CAS-Nummer: 38289-29-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...