trans-4-Propylcyclohexancarbonsäure
Lieferant: COMBI-BLOCKS
CA-4500-5G
CA-4500-25G
CA-4500-100G
COBBCA-4500-5GEA
0
EUR
COBBCA-4500-5G
COBBCA-4500-25G
COBBCA-4500-100G
trans-4-Propylcyclohexancarbonsäure
trans-4-Propylcyclohexanecarboxylic acid
Formel:
C₁₀H₁₈O₂ |
MDL:
MFCD00059559 CAS-Nummer: 38289-27-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...