Methyl-4-fluor-2-benzo[b]thiophencarboxylat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 4-fluorobenzo[b]thiophene-2-carboxylate

4T-1304.1 4T-1304.10 4T-1304.5 4T-1304.500
BIOT4T-1304.1EA 0 EUR
BIOT4T-1304.1 BIOT4T-1304.10 BIOT4T-1304.5 BIOT4T-1304.500
Methyl-4-fluor-2-benzo[b]thiophencarboxylat
Methyl-4-fluor-2-benzo[b]thiophencarboxylat
Formel: C₁₀H₇FO₂S
MDL: MFCD06200964
CAS-Nummer: 220180-55-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR