3',4'-(Methylendioxy)acetophenon ≥98%
Lieferant: Thermo Scientific
A13597.09
A13597.18
A13597.30
A13597.09EA
41
EUR
A13597.09
A13597.18
A13597.30
3',4'-(Methylendioxy)acetophenon ≥98%
3',4'-(Methylendioxy)acetophenon
Purity: 98%
CAS No.: 3162-29-6
Molecular Formula: C9H8O3
Formula Weight: 164.16
Melting Point: 85-89°
Boiling Point: 152-155°/15mm
MDL No.: MFCD00005831
Formel:
C₉H₈O₃ Molecular Weight: 164,16 g/mol Siedepunkt: 152…155 °C (15 mmHg) Schmelzpunkt: 85…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005831 CAS-Nummer: 3162-29-6 EINECS: 221-613-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...