1-(2-Aminoethyl)pyrrolidin 99%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
N-(2-Aminoethyl)pyrrolidine, Pyrrolidinoethylamine
B24645.06
B24645.14
B24645.22
B24645.06EA
50.8
EUR
B24645.06
B24645.14
B24645.22
1-(2-Aminoethyl)pyrrolidin 99%
1-(2-Aminoethyl)pyrrolidin
Purity: 99%
CAS No.: 7154-73-6
Molecular Formula: C6H14N2
Formula Weight: 114.19
Flash Point: 47°(116°F)
Refractive Index: 1.4687
Odour: Amine-like
Boiling Point: 161°
Beilstein Reference: 102978
UN No.: 2734
MDL No.: MFCD00003182
Formel:
C₆H₁₄N₂ Molecular Weight: 114.19 g/mol Siedepunkt: 161 °C Dichte: 0.904 Flammpunkt: 47 °C (116 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00003182 CAS-Nummer: 7154-73-6 EINECS: 230-509-5 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...