Bis(α,α-dimethylbenzyl)peroxid (Dicumylperoxid), Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
Bis(1-methyl-1-phenylethyl)peroxide, Bis(alpha,alpha-dimethylbenzyl)peroxide, Dicumyl Peroxide
M-Clarity™ Qualitätsstufe = MQ200
329541-100G
329541-500G
SIAL329541-100GEA
89.5
EUR
SIAL329541-100G
SIAL329541-500G
Bis(α,α-dimethylbenzyl)peroxid (Dicumylperoxid), Sigma-Aldrich®
Bis(α,α-dimethylbenzyl)peroxid (Dicumylperoxid)
Formel:
C₁₈H₂₂O₂ Molecular Weight: 270.37 g/mol |
MDL:
MFCD00036227 CAS-Nummer: 80-43-3 EINECS: 201-279-3 UN: 3110 ADR: 5.2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...