4-Brom-DL-phenylalanin

Lieferant: BACHEM BIOCHEMICA

4007001.0005 4007001.0025
BACE4007001.0005EA 0 EUR
BACE4007001.0005 BACE4007001.0025
4-Brom-DL-phenylalanin
4-Brom-DL-phenylalanin
Formel: C₉H₁₀BrNO₂
Molecular Weight: 244.09 g/mol
MDL: MFCD00002599
CAS-Nummer: 14091-15-7
EINECS: 237-941-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR