4-Chlor-DL-phenylalanin

Lieferant: BACHEM BIOCHEMICA

Synonyme: p-Chloro-DL-Phenylalanine, H-DL-Phe(4-Cl)-OH

4002988.0025 4002988.0100
BACE4002988.0025EA 0 EUR
BACE4002988.0025 BACE4002988.0100
4-Chlor-DL-phenylalanin
4-Chlor-DL-phenylalanin
Formel: C₉H₁₀ClNO₂
MDL: MFCD00002601
CAS-Nummer: 7424-00-2
EINECS: 231-051-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR