4-Chlor-DL-phenylalanin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: p-Chloro-DL-Phenylalanine, H-DL-Phe(4-Cl)-OH

M-Clarity™ Qualitätsstufe = MQ100

C6506-25G C6506-5G
SIALC6506-25GEA 291 EUR
SIALC6506-25G SIALC6506-5G
4-Chlor-DL-phenylalanin, Sigma-Aldrich®
4-Chlor-DL-phenylalanin
Formel: C₉H₁₀ClNO₂
Molecular Weight: 199,6 g/mol
MDL: MFCD00002601
CAS-Nummer: 7424-00-2
EINECS: 231-051-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR