3-(3,4-Dihydroxyphenyl)-DL-alanin ≥98%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
DL-DOPA
41535.06
41535.14
41535.06EA
46.5
EUR
41535.06
41535.14
3-(3,4-Dihydroxyphenyl)-DL-alanin ≥98%
3-(3,4-Dihydroxyphenyl)-DL-alanin
Purity: 98%
CAS No.: 63-84-3
Molecular Formula: C9H11NO4
Formula Weight: 197.19
Physical Form: Crystalline
Odour: Odourless
Merck Reference: 14,3420
MDL No.: MFCD00063060
Formel:
C₉H₁₁NO₄ Molecular Weight: 197.19 g/mol Schmelzpunkt: 290 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063060 CAS-Nummer: 63-84-3 EINECS: 200-566-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...