4-Fluor-DL-phenylalanin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

F5251-1G F5251-5G
SIALF5251-1GEA 111 EUR
SIALF5251-1G SIALF5251-5G
4-Fluor-DL-phenylalanin, Sigma-Aldrich®
4-Fluor-DL-phenylalanin
Formel: C₉H₁₀FNO₂
Molecular Weight: 183,18 g/mol
MDL: MFCD00002600
CAS-Nummer: 51-65-0
EINECS: 200-113-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR