4-Chlor-L-phenylalanin, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
L-4-Chlorophenylalanine, H-Phe(4-Cl)-OH
M-Clarity™ Qualitätsstufe = MQ200
C8655-1G
C8655-250MG
SIALC8655-1GEA
325
EUR
SIALC8655-1G
SIALC8655-250MG
4-Chlor-L-phenylalanin, Sigma-Aldrich®
4-Chlor-L-phenylalanin
Formel:
C₉H₁₀ClNO₂ |
MDL:
MFCD00063065 CAS-Nummer: 14173-39-8 EINECS: 238-023-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...