3-(3,4-Dihydroxyphenyl)-D-alanin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

D9378-1G D9378-250MG
SIALD9378-1GEA 771 EUR
SIALD9378-1G SIALD9378-250MG
3-(3,4-Dihydroxyphenyl)-D-alanin, Sigma-Aldrich®
3-(3,4-Dihydroxyphenyl)-D-alanin
Formel: C₉H₁₁NO₄
CAS-Nummer: 5796-17-8
EINECS: 227-343-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR