3-(3,4-Dihydroxyphenyl)-L-alanin

Lieferant: Cayman Chemical

Synonyme: Dopar, L-dopa, L-β-3,4-Dihydroxyphenyl-Alanine, 3-Hydroxy-L-tyrosine, Levodopa

13248-5 13248-25 13248-50 13248-100
CAYM13248-5EA 26 EUR
CAYM13248-5 CAYM13248-25 CAYM13248-50 CAYM13248-100
3-(3,4-Dihydroxyphenyl)-L-alanin
3-(3,4-Dihydroxyphenyl)-L-alanin
Formel: C₉H₁₁NO₄
Molecular Weight: 197.19 g/mol
MDL: MFCD00002598
CAS-Nummer: 59-92-7
EINECS: 200-445-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR