4-Fluor-L-phenylalanin 98
Lieferant: Thermo Fisher Scientific
Synonyme:
(S)-2-Amino-3-(4-fluorophenyl)propanoic acid, H-Phe(4-F)-OH
L19934.03
L19934.06
L19934.03EA
200
EUR
L19934.03
L19934.06
4-Fluor-L-phenylalanin 98
4-Fluor-L-phenylalanin
Purity: 98+%
CAS No.: 1132-68-9
Molecular Formula: C9H10FNO2
Formula Weight: 183.18
Melting Point: ca 240° dec.
Beilstein Reference: 2416148
Formel:
C₉H₁₀FNO₂ Molecular Weight: 183.2 g/mol Schmelzpunkt: ∼ 240 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00063064 CAS-Nummer: 1132-68-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...