4-Fluor-L-phenylalanin

Lieferant: BACHEM BIOCHEMICA

Synonyme: (S)-2-Amino-3-(4-fluorophenyl)propanoic acid, H-Phe(4-F)-OH

4007214.0001 4007214.0005
BACE4007214.0001EA 0 EUR
BACE4007214.0001 BACE4007214.0005
4-Fluor-L-phenylalanin
4-Fluor-L-phenylalanin
Formel: C₉H₁₀FNO₂
Molecular Weight: 183.2 g/mol
MDL: MFCD00063064
CAS-Nummer: 1132-68-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR