4-Iod-L-phenylalanin 95%
Lieferant: Thermo Fisher Scientific
Synonyme:
H-Phe(4-I)-OH
H51979.03
H51979.06
H51979.03EA
135
EUR
H51979.03
H51979.06
4-Iod-L-phenylalanin 95%
4-Iod-L-phenylalanin
Purity: 95%
CAS No.: 24250-85-9
Molecular Formula: C9H10INO2
Formula Weight: 291.09
Beilstein Reference: 4411317
Optical Rotation: -6.5° (c=1 in acetic acid:water (4:1))
Formel:
C₉H₁₀INO₂ Molecular Weight: 291.1 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002602 CAS-Nummer: 24250-85-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...