4-(1H-Imidazol-1-yl)phenol ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-(1-Imidazolyl)phenol, 4-(Imidazol-1-yl)phenol

A13211.03 A13211.06 A13211.14
A13211.03EA 25.7 EUR
A13211.03 A13211.06 A13211.14
4-(1H-Imidazol-1-yl)phenol ≥97%
4-(1H-Imidazol-1-yl)phenol

Purity: 97% CAS No.: 10041-02-8 Molecular Formula: C9H8N2O Formula Weight: 160.18 Melting Point: 204-208° Beilstein Reference: 509999

Formel: C₉H₈N₂O
Molecular Weight: 160,18 g/mol
Schmelzpunkt: 204…208 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005281
CAS-Nummer: 10041-02-8
EINECS: 233-121-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR