4-Amino-2-chlorpyrimidin-5-carbosäurenitril

Lieferant: BIONET RESEARCH

Synonyme: 4-Amino-2-chloro-5-pyrimidinecarbonitrile

5Y-5028.1 5Y-5028.5 5Y-5028.500
BIOT5Y-5028.1EA 0 EUR
BIOT5Y-5028.1 BIOT5Y-5028.5 BIOT5Y-5028.500
4-Amino-2-chlorpyrimidin-5-carbosäurenitril
4-Amino-2-chlorpyrimidin-5-carbosäurenitril
Formel: C₅H₃ClN₄
Molecular Weight: 154,56 g/mol
MDL: MFCD00052343
CAS-Nummer: 94741-69-2
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR