4-Aminopyrimidin-5-carbosäurenitril
Lieferant: CHEMPUR
SEW03364.250
SEW03364.1
CHMPSEW03364.250EA
0
EUR
CHMPSEW03364.250
MAYBSEW03364.1
4-Aminopyrimidin-5-carbosäurenitril
4-Aminopyrimidin-5-carbosäurenitril
Formel:
C₅H₄N₄ Molecular Weight: 120,11 g/mol |
MDL:
MFCD00054552 CAS-Nummer: 16357-69-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...