2,4,5-Triamino-6-hydroxypyrimidinsulfat 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2,5,6-Triaminopyrimidin-4(1H)-one sulphate, 4-Hydroxy-2,5,6-triaminopyrimidine sulphate, 6-Hydroxy-2,4,5-triaminopyrimidine sulphate

B21989.14 B21989.22
B21989.14EA 43.5 EUR
B21989.14 B21989.22
2,4,5-Triamino-6-hydroxypyrimidinsulfat 97%
2,4,5-Triamino-6-hydroxypyrimidinsulfat

Purity: 97% CAS No.: 35011-47-3 Molecular Formula: C4H7N5O.H2SO4 Formula Weight: 239.21 Melting Point: 300° Beilstein Reference: 4621048

Formel: C₄H₉N₅O₅S
Molecular Weight: 239,21 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012785
CAS-Nummer: 39267-74-8
EINECS: 254-393-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR