4-Amino-2-mercaptopyrimidin
Lieferant: Thermo Scientific
Synonyme:
4-Amino-2-mercaptopyrimidine, 4-Aminopyrimidine-2-thiol, 4-Amino-2-sulphanylpyrimidine
AC21477.1
AC21477.10
MAYBAC21477.1EA
0
EUR
MAYBAC21477.1
MAYBAC21477.10
4-Amino-2-mercaptopyrimidin
4-Amino-2-mercaptopyrimidine
Formel:
C₄H₅N₃S Molecular Weight: 127,1676 g/mol |
MDL:
MFCD00006075 CAS-Nummer: 333-49-3 EINECS: 206-374-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...