1-(2-Aminophenyl)pyrrol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
2-(1H-Pyrrol-1-yl)aniline
M-Clarity™ Qualitätsstufe = MQ100
196940-10G
196940-1G
SIAL196940-10GEA
199
EUR
SIAL196940-10G
SIAL196940-1G
1-(2-Aminophenyl)pyrrol, Sigma-Aldrich®
1-(2-Aminophenyl)pyrrole
Formel:
C₁₀H₁₀N₂ Molecular Weight: 158.20 g/mol |
MDL:
MFCD00005344 CAS-Nummer: 6025-60-1 EINECS: 227-884-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...