2-Methyl-3-(pyrrol-1-yl)anilin

Lieferant: BIONET RESEARCH

1W-0835.500 1W-0835.1 1W-0835.5 1W-0835.10
BIOT1W-0835.500EA 0 EUR
BIOT1W-0835.500 BIOT1W-0835.1 BIOT1W-0835.5 BIOT1W-0835.10
2-Methyl-3-(pyrrol-1-yl)anilin
2-Methyl-3-(pyrrol-1-yl)anilin
Formel: C₁₁H₁₂N₂
CAS-Nummer: 137352-75-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR