5-Chlor-2-benzofurancarbonsäure ≥97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 5-Chlorobenzo[b]furan-2-carboxylic acid

H50239.03 H50239.06
H50239.03EA 108 EUR
H50239.03 H50239.06
5-Chlor-2-benzofurancarbonsäure ≥97%
5-Chlorobenzofuran-2-carboxylic acid

Purity: 97% CAS No.: 10242-10-1 Molecular Formula: C9H5ClO3 Formula Weight: 196.59 MDL No.: MFCD00060512

Formel: C₉H₅ClO₃
Molecular Weight: 196,59 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00060512
CAS-Nummer: 10242-10-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR