Ethyl-7-methoxy-2-benzofurancarboxylat ≥97%
Lieferant: Thermo Scientific
Synonyme:
Ethyl 7-methoxybenzofuran-2-carboxylate
B21066.03
B21066.06
B21066.14
B21066.03EA
22
EUR
B21066.03
B21066.06
B21066.14
Ethyl-7-methoxy-2-benzofurancarboxylat ≥97%
Ethyl-7-methoxy-2-benzofurancarboxylat
Purity: 97%
CAS No.: 50551-58-1
Molecular Formula: C12H12O4
Formula Weight: 220.22
Melting Point: 83-86°
MDL No.: MFCD00060510
Formel:
C₁₂H₁₂O₄ Molecular Weight: 220,22 g/mol Schmelzpunkt: 83…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00060510 CAS-Nummer: 50551-58-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...