Dibenzothiophen-4-boronsäure

Lieferant: COMBI-BLOCKS

Synonyme: 4-Dibenzothiopheneboronic acid

BB-3277-5G BB-3277-1G BB-3277-25G BB-3277-100G
COBBBB-3277-5GEA 50.8 EUR
COBBBB-3277-5G COBBBB-3277-1G COBBBB-3277-25G COBBBB-3277-100G
Dibenzothiophen-4-boronsäure
Dibenzothiophen-4-boronsäure
Formel: C₁₂H₉BO₂S
Molecular Weight: 228,08 g/mol
MDL: MFCD01318182
CAS-Nummer: 108847-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR