Dibenzothiophen-4-boronsäure

Lieferant: FLUOROCHEM

Synonyme: 4-Dibenzothiopheneboronic acid

011027-1G 011027-5G
FLUO011027-1GEA 29.9 EUR
FLUO011027-1G FLUO011027-5G
Dibenzothiophen-4-boronsäure
Dibenzothiophen-4-boronsäure
Formel: C₁₂H₉BO₂S
Molecular Weight: 228,08 g/mol
MDL: MFCD01318182
CAS-Nummer: 108847-20-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR