Ethylsuccinylchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Ethyl 4-chloro-4-oxobutanoate

M-Clarity™ Qualitätsstufe = MQ100

190705-100G 190705-25G
SIAL190705-100GEA 134 EUR
SIAL190705-100G SIAL190705-25G
Ethylsuccinylchlorid, Sigma-Aldrich®
Ethylsuccinylchlorid
Formel: C₆H₉ClO₃
Molecular Weight: 164,59 g/mol
MDL: MFCD00000751
CAS-Nummer: 14794-31-1
EINECS: 238-855-9
UN: 3265
ADR: 8,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR