1-Brom-5-iod-4-nitro-2-(trifluormethoxy)benzol
Lieferant: COMBI-BLOCKS
HC-2984-5G
HC-2984-1G
COBBHC-2984-5GEA
0
EUR
COBBHC-2984-5G
COBBHC-2984-1G
1-Brom-5-iod-4-nitro-2-(trifluormethoxy)benzol
1-Brom-5-iod-4-nitro-2-(trifluormethoxy)benzol
Formel:
C₇H₂BrF₃INO₃ |
MDL:
MFCD12913963 CAS-Nummer: 1187386-19-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...