Monomethylterephthalat ≥99%

Lieferant: Thermo Scientific
Achtung

Synonyme: Methyl hydrogen terephthalate, 4-(Methoxycarbonyl)benzoic acid

H54391.06 H54391.22 H54391.14
H54391.06EA 46.7 EUR
H54391.06 H54391.22 H54391.14
Monomethylterephthalat ≥99%
Monomethylterephthalat

Purity: 99+% CAS No.: 1679-64-7 Molecular Formula: C9H8O4 Formula Weight: 180.16 Melting Point: 220-223° Beilstein Reference: 1911082

Formel: C₉H₈O₄
Molecular Weight: 180,16 g/mol
Schmelzpunkt: 220…223 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002557
CAS-Nummer: 1679-64-7
EINECS: 216-849-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR