Monomethylterephthalat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Methyl hydrogen terephthalate, 4-(Methoxycarbonyl)benzoic acid

M-Clarity™ Qualitätsstufe = MQ100

328383-25G 328383-5G
SIAL328383-25GEA 105 EUR
SIAL328383-25G SIAL328383-5G
Monomethylterephthalat, Sigma-Aldrich®
Monomethylterephthalat
Formel: C₉H₈O₄
Molecular Weight: 180,16 g/mol
MDL: MFCD00002557
CAS-Nummer: 1679-64-7
EINECS: 216-849-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR