(Bis(trifluoracetoxy)iod)benzol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
[Bis(trifluoroacetoxy)](phenyl)iodane
M-Clarity™ Qualitätsstufe = MQ200
15230-10G
15230-50G
SIAL15230-10GEA
81.4
EUR
SIAL15230-10G
SIAL15230-50G
(Bis(trifluoracetoxy)iod)benzol, Sigma-Aldrich®
(Bis(trifluoracetoxy)iod)benzol
Formel:
C₁₀H₅F₆IO₄ Molecular Weight: 430,04 g/mol |
MDL:
MFCD00009672 CAS-Nummer: 2712-78-9 EINECS: 220-308-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...