4'-(Dimethylamino)acetophenon

Lieferant: COMBI-BLOCKS

Synonyme: 1-(4-(Dimethylamino)phenyl)ethanone

OT-0676-5G OT-0676-25G OT-0676-100G
COBBOT-0676-5GEA 38.8 EUR
COBBOT-0676-5G COBBOT-0676-25G COBBOT-0676-100G
4'-(Dimethylamino)acetophenon
4'-(Dimethylamino)acetophenon
Formel: C₁₀H₁₃NO
Molecular Weight: 163,2200 g/mol
MDL: MFCD00017246
CAS-Nummer: 2124-31-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR